2,3,4-Trichlornitrobenzol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Nitro-2,3,4-trichlorobenzene, 1,2,3-Trichloro-4-nitrobenzene, 4-Nitro-1,2,3-trichlorobenzene
M-Clarity™ Qualitätsstufe = MQ100
T55158-10G
SIALT55158-10GEA
39.6
EUR
SIALT55158-10G
2,3,4-Trichlornitrobenzol, Sigma-Aldrich®
2,3,4-Trichlornitrobenzol
Formel:
C₆H₂Cl₃NO₂ Molecular Weight: 226,45 g/mol Schmelzpunkt: 55,5 °C |
MDL:
MFCD00007063 CAS-Nummer: 17700-09-3 EINECS: 241-705-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...