Tris(hydroxymethyl)aminomethan (TRIS, Trometamol), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Aminomethylidintrimethanol, Tris(hydroxymethyl)methylamin, THAM, 2-Amino-2-(hydroxymethyl)propan-1,3-diol, Tromethamin, TRIS Base, 2-Amino-2-(hydroxymethyl)-1,3-propandiol
M-Clarity™ Qualitätsstufe = MQ200
T6687-100G
T6687-10KG
T6687-25KG
T6687-5KG
SIALT6687-100GEA
121
EUR
SIALT6687-100G
SIALT6687-10KG
SIALT6687-25KG
SIALT6687-5KG
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol), Sigma-Aldrich®
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Formel:
NH₂C(CH₂OH)₃ Molecular Weight: 121.14 g/mol Siedepunkt: 357 Schmelzpunkt: 169…172 °C Dichte: 1.328 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004679 CAS-Nummer: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...