Tetracyclin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

T7660-5G
SIALT7660-5GEA 63.4 EUR
SIALT7660-5G
Tetracyclin Hydrochlorid, Sigma-Aldrich®
Tetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₈
Molecular Weight: 480.9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078142
CAS-Nummer: 64-75-5
EINECS: 200-593-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR