2,3,3-Trimethylindolenin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
T76805-25G
SIALT76805-25GEA
75.3
EUR
SIALT76805-25G
2,3,3-Trimethylindolenin, Sigma-Aldrich®
2,3,3-Trimethylindolenin
Formel:
C₁₁H₁₃N Molecular Weight: 159,23 g/mol |
MDL:
MFCD00005724 CAS-Nummer: 1640-39-7 EINECS: 216-685-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...