D-Threonin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

T8250-100G T8250-25G T8250-5G
SIALT8250-100GEA 948 EUR
SIALT8250-100G SIALT8250-25G SIALT8250-5G
D-Threonin, Sigma-Aldrich®
D-Threonin
Formel: C₄H₉NO₃
MDL: MFCD00064269
CAS-Nummer: 632-20-2
EINECS: 211-171-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR