DL-Threonin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
T8375-100G
SIALT8375-100GEA
315
EUR
SIALT8375-100G
DL-Threonin, Sigma-Aldrich®
DL-Threonin
Formel:
C₄H₉NO₃ Molecular Weight: 119,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063722 CAS-Nummer: 80-68-2 EINECS: 201-300-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...