3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydrochloride TMB, TMB-HCl

M-Clarity™ Qualitätsstufe = MQ200

T8768-100MG T8768-1G T8768-5G
SIALT8768-100MGEA 27.1 EUR
SIALT8768-100MG SIALT8768-1G SIALT8768-5G
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel: C₁₆H₂₀N₂·2HCl·xH₂O
Molecular Weight: 313.27 g/mol (anhy)
MDL: MFCD00150104
CAS-Nummer: 207738-08-7
EINECS: 264-769-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR