3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dihydrochloride TMB, TMB-HCl
M-Clarity™ Qualitätsstufe = MQ200
T8768-100MG
T8768-1G
T8768-5G
SIALT8768-100MGEA
27.1
EUR
SIALT8768-100MG
SIALT8768-1G
SIALT8768-5G
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel:
C₁₆H₂₀N₂·2HCl·xH₂O Molecular Weight: 313.27 g/mol (anhy) |
MDL:
MFCD00150104 CAS-Nummer: 207738-08-7 EINECS: 264-769-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...