Thymidin (2'-Deoxythymidin), Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

M-Clarity™ Qualitätsstufe = MQ200

T9250-1G T9250-5G T9250-10G T9250-25G
SIALT9250-1GEA 77.5 EUR
SIALT9250-1G SIALT9250-5G SIALT9250-10G SIALT9250-25G
Thymidin (2'-Deoxythymidin), Sigma-Aldrich®
Thymidin (2'-Deoxythymidin)
Formel: C₁₀H₁₄N₂O₅
Molecular Weight: 242.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006537
CAS-Nummer: 50-89-5
EINECS: 200-070-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR