TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propansulfonsäure), Sigma-Aldrich®

Lieferant: Merck

Synonyme: TAPSO, 2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulphonic acid, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulphonic acid

M-Clarity™ Qualitätsstufe = MQ200

T9269-100G
SIALT9269-100GEA 152 EUR
SIALT9269-100G
TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propansulfonsäure), Sigma-Aldrich®
TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propansulfonsäure)
Formel: C₇H₁₇NO₇S
Molecular Weight: 259,28 g/mol
MDL: MFCD00038352
CAS-Nummer: 68399-81-5
EINECS: 269-993-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR