Triethanolamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

T9534-100G T9534-1KG T9534-250G T9534-500G
SIALT9534-100GEA 182 EUR
SIALT9534-100G SIALT9534-1KG SIALT9534-250G SIALT9534-500G
Triethanolamin Hydrochlorid, Sigma-Aldrich®
Triethanolamin Hydrochlorid
Formel: C₆H₁₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00012596
CAS-Nummer: 637-39-8
EINECS: 211-284-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR