D(-)-Valin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(-)-Valin, (R)-(-)-2-Amino-3-methylbutansäure, (R)-(-)-2-Aminoisovaleriansäure, H-D-Val-OH
MQ200
V1255-1G
V1255-25G
V1255-5G
SIALV1255-1GEA
40.2
EUR
SIALV1255-1G
SIALV1255-25G
SIALV1255-5G
D(-)-Valin, Sigma-Aldrich®
D(-)-Valin
Formel:
C₅H₁₁NO₂ Molecular Weight: 117.15 g/mol Siedepunkt: 217 Schmelzpunkt: 86 |
MDL:
MFCD00064219 CAS-Nummer: 640-68-6 EINECS: 211-368-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...