Isopentylpropionat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Isoamyl Propionate, Propionic acid isoamyl ester
M-Clarity™ Qualitätsstufe = MQ300
W208213-1KG-K
W208213-4KG-K
SIALW208213-1KG-KEA
450
EUR
SIALW208213-1KG-K
SIALW208213-4KG-K
Isopentylpropionat, Sigma-Aldrich®
Isopentylpropionat
Formel:
C₈H₁₆O₂ |
MDL:
MFCD00048611 CAS-Nummer: 105-68-0 EINECS: 203-322-1 UN: 3272 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...