(±)-γ-Dodecalacton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-γ-Dodekalacton, (±)-Dodecan-4-olid, (±)-4-Hydroxydodecansäurelacton, (±)-Dihydro-5-octylfuran-2(3H)-on

MQ100

W240001-100G-K W240001-1KG-K W240001-5KG-K
SIALW240001-100G-KEA 89.8 EUR
SIALW240001-100G-K SIALW240001-1KG-K SIALW240001-5KG-K
(±)-γ-Dodecalacton, Sigma-Aldrich®
(±)-γ-Dodecalacton
Formel: C₁₂H₂₂O₂
Molecular Weight: 198.31 g/mol
MDL: MFCD00036499
CAS-Nummer: 2305-05-7
EINECS: 218-971-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR