Geraniol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ300
W250708-1KG-K
W250708-20KG-K
W250708-9KG-K
SIALW250708-1KG-KEA
89.8
EUR
SIALW250708-1KG-K
SIALW250708-20KG-K
SIALW250708-9KG-K
Geraniol, Sigma-Aldrich®
Geraniol
Formel:
C₁₀H₁₈O |
MDL:
MFCD00002917 CAS-Nummer: 106-24-1 EINECS: 203-377-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...