Geraniol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ300
W250716-1KG-K
W250716-4KG-K
W250716-9KG-K
SIALW250716-1KG-KEA
199
EUR
SIALW250716-1KG-K
SIALW250716-4KG-K
SIALW250716-9KG-K
Geraniol, Sigma-Aldrich®
Geraniol
Formel:
C₁₀H₁₈O |
MDL:
MFCD00002917 CAS-Nummer: 106-24-1 EINECS: 203-377-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...