Geranylacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-3,7-Dimethylocta-2,6-dien-1-yl acetate

M-Clarity™ Qualitätsstufe = MQ300

W250902-10KG-K
SIALW250902-10KG-KEA 545 EUR
SIALW250902-10KG-K
Geranylacetat, Sigma-Aldrich®
Geranylacetat
Formel: C₁₂H₂₀O₂
Molecular Weight: 196,29 g/mol
MDL: MFCD00015037
CAS-Nummer: 105-87-3
EINECS: 203-341-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR