Geranylacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W250910-100G-K W250910-1KG-K W250910-5KG-K
SIALW250910-100G-KEA 90 EUR
SIALW250910-100G-K SIALW250910-1KG-K SIALW250910-5KG-K
Geranylacetat, Sigma-Aldrich®
Geranylacetat
Formel: C₁₂H₂₀O₂
MDL: MFCD00015037
CAS-Nummer: 105-87-3
EINECS: 203-341-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR