4-Methyl-1-phenyl-2-pentanon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-1-phenylpentan-2-one

M-Clarity™ Qualitätsstufe = MQ300

W274003-100G-K
SIALW274003-100G-KEA 636 EUR
SIALW274003-100G-K
4-Methyl-1-phenyl-2-pentanon, Sigma-Aldrich®
4-Methyl-1-phenyl-2-pentanon
Formel: (C₆H₅)CH₂C(O)CH₂CH(CH₃)₂
Molecular Weight: 176,26 g/mol
MDL: MFCD00015639
CAS-Nummer: 5349-62-2
EINECS: 226-316-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR