Polysorbat 20, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ300
W291501-10KG
W291501-1KG
W291501-25KG
SIALW291501-10KGEA
268
EUR
SIALW291501-10KG
SIALW291501-1KG
SIALW291501-25KG
Polysorbat 20, Sigma-Aldrich®
Polysorbat 20
Formel:
C₅₈H₁₁₄O₂₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00165986 CAS-Nummer: 9005-64-5 EINECS: 500-018-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...