(±)-α-Terpineol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

M-Clarity™ Qualitätsstufe = MQ100

W304522-10KG-K W304522-1KG-K W304522-5KG-K
SIALW304522-10KG-KEA 930 EUR
SIALW304522-10KG-K SIALW304522-1KG-K SIALW304522-5KG-K
(±)-α-Terpineol, Sigma-Aldrich®
(±)-α-Terpineol
Formel: C₁₀H₁₈O
Molecular Weight: 154.25 g/mol
MDL: MFCD00001557
CAS-Nummer: 98-55-5
EINECS: 202-680-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR