5-Methylthiophen-2-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W320900-100G-K W320900-1KG-K
SIALW320900-100G-KEA 253 EUR
SIALW320900-100G-K SIALW320900-1KG-K
5-Methylthiophen-2-carbaldehyd, Sigma-Aldrich®
5-Methylthiophen-2-carbaldehyd
Formel: C₆H₆OS
MDL: MFCD00005434
CAS-Nummer: 13679-70-4
EINECS: 237-178-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR