α-Angelicalacton, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Methylfuran-2(3H)-on
M-Clarity™ Qualitätsstufe = MQ300
W329304-100G-K
W329304-1KG-K
W329304-250G-K
SIALW329304-100G-KEA
226
EUR
SIALW329304-100G-K
SIALW329304-1KG-K
SIALW329304-250G-K
α-Angelicalacton, Sigma-Aldrich®
α-Angelicalacton
Formel:
C₅H₆O₂ Molecular Weight: 98.1 g/mol Siedepunkt: 249 Schmelzpunkt: 0 Dichte: 1.0904 g/cm³ (12 °C) |
MDL:
MFCD00005375 CAS-Nummer: 591-12-8 EINECS: 209-701-8 Merck Index: 12,00686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...