3-(Methylsulfanyl)butanal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Methylthio)butyraldehyde

M-Clarity™ Qualitätsstufe = MQ300

W337404-100G-K W337404-1KG-K W337404-250G-K
SIALW337404-100G-KEA 276 EUR
SIALW337404-100G-K SIALW337404-1KG-K SIALW337404-250G-K
3-(Methylsulfanyl)butanal, Sigma-Aldrich®
3-(Methylthio)butanal
Formel: C₅H₁₀OS
Molecular Weight: 118.2 g/mol
MDL: MFCD00036622
CAS-Nummer: 16630-52-7
EINECS: 240-678-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR