2-sec-Butyl-3-methoxypyrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(sec-Butyl)-3-methoxypyrazine

M-Clarity™ Qualitätsstufe = MQ300

W343307-100G-K W343307-1KG-K
SIALW343307-100G-KEA 707 EUR
SIALW343307-100G-K SIALW343307-1KG-K
2-sec-Butyl-3-methoxypyrazin, Sigma-Aldrich®
2-sec-Butyl-3-methoxypyrazin
Formel: C₉H₁₄N₂O
Molecular Weight: 166.2203 g/mol
MDL: MFCD00006136
CAS-Nummer: 24168-70-5
EINECS: 246-050-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR