2-Methylbutylisovalerat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isovaleriansäure(2-methylbutyl)ester, 2-Methylbutylisopentanoat, 3-Methylbuttersäure(2-methylbutyl)ester, Isopropylessigsäure(2-methylbutyl)ester, 3-Methylbutansäure(2-methylbutyl)ester

M-Clarity™ Qualitätsstufe = MQ300

W350613-1KG-K
SIALW350613-1KG-KEA 306 EUR
SIALW350613-1KG-K
2-Methylbutylisovalerat, Sigma-Aldrich®
2-Methylbutylisovalerat
Formel: C₁₀H₂₀O₂
Molecular Weight: 172,27 g/mol
CAS-Nummer: 2445-77-4
EINECS: 219-496-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR