2,6-Dimethoxy-p-kresol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dimethoxy-4-hydroxytoluol, 2,6-Dimethoxy-4-methylphenol, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzol

M-Clarity™ Qualitätsstufe = MQ300

W370401-1KG W370401-25G
SIALW370401-1KGEA 2920 EUR
SIALW370401-1KG SIALW370401-25G
2,6-Dimethoxy-p-kresol, Sigma-Aldrich®
2,6-Dimethoxy-p-kresol
Formel: C₉H₁₂O₃
Molecular Weight: 168.19 g/mol
MDL: MFCD00017289
CAS-Nummer: 6638-05-7
EINECS: 229-641-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR