2-Methylchinoxalin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
MQ300
W511609-1KG
W511609-5KG
SIALW511609-1KGEA
1180
EUR
SIALW511609-1KG
SIALW511609-5KG
2-Methylchinoxalin, Sigma-Aldrich®
2-Methylquinoxaline
Formel:
C₉H₈N₂ Molecular Weight: 144.18 g/mol Siedepunkt: 245 Dichte: 1.11 g/cm³ (20 °C) |
MDL:
MFCD00006727 CAS-Nummer: 7251-61-8 EINECS: 230-664-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...