Cumarin ≥98%, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 1,2-Benzopyron

M-Clarity™ Qualitätsstufe = MQ300

W526509-10KG
SIALW526509-10KGEA 473 EUR
SIALW526509-10KG
Cumarin ≥98%, Sigma-Aldrich®
Cumarin

Coumarin is an aroma compound responsible for the characteristic odor of hay. It is one of the major volatile compounds found in the aerial parts of bastard balm.


  • biological source: synthetic
  • grade: Halal

Formel: C₉H₆O₂
Molecular Weight: 146,15 g/mol
Siedepunkt: 298 °C (Lit.)
Schmelzpunkt: 67…73 °C (Lit.)
Dichte: 0,935 g/cm³ (20 °C)
Flammpunkt: 162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006850
CAS-Nummer: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR