Trimethylamin 25% (w/w) in Propylenglykol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W550132-1KG
SIALW550132-1KGEA 710 EUR
SIALW550132-1KG
Trimethylamin 25% (w/w) in Propylenglykol, Sigma-Aldrich®
Trimethylamin
Formel: C₃H₉N
Molecular Weight: 59,11 g/mol
MDL: MFCD00008327
CAS-Nummer: 75-50-3
EINECS: 200-875-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR