Laboratory Brushes
Lieferant: Merck
Z281344-1PAK
SIALZ281344-1PAKEA 45.2 EUR
SIALZ281344-1PAK
Laboratory Brushes
Bürsten
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water.
  • FDA and USDA approved materials