Allyltriphenylsilan

Lieferant: CHEMPUR SILANE

SILS00475.5 SILS00475.25
SILNSILS00475.5EA 0 EUR
SILNSILS00475.5 SILNSILS00475.25
Allyltriphenylsilan
Allyltriphenylsilan
Formel: H₂CCHCH₂Si(C₆H₅)₃
MDL: MFCD00077644
CAS-Nummer: 18752-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR