1,4-Bis(trimethylsilyl)benzol
Lieferant: CHEMPUR SILANE
Synonyme:
1,4-Phenylenbis(trimethylsilan)
SILS02500.5
SILS02500.10
SILS02500.50
SILNSILS02500.5EA
0
EUR
SILNSILS02500.5
SILNSILS02500.10
SILNSILS02500.50
1,4-Bis(trimethylsilyl)benzol
1,4-Bis(trimethylsilyl)benzol
Formel:
C₁₂H₂₂Si₂ Siedepunkt: 83 °C (0,5 mmHg) Schmelzpunkt: 97,5 °C |
MDL:
MFCD00015592 CAS-Nummer: 13183-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...