1,4-Bis(trimethylsilyl)-1,3-butadiin

Lieferant: CHEMPUR SILANE

SILS02525.1 SILS02525.10 SILS02525.25
SILNSILS02525.1EA 0 EUR
SILNSILS02525.1 SILNSILS02525.10 SILNSILS02525.25
1,4-Bis(trimethylsilyl)-1,3-butadiin
1,4-Bis(trimethylsilyl)-1,3-butadiin
Formel: C₁₀H₁₈Si₂
MDL: MFCD00009839
CAS-Nummer: 4526-07-2
UN: 1325
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR