Bis(trimethylsilyl)methan

Lieferant: CHEMPUR SILANE

SILS02725.1 SILS02725.5 SILS02725.25
SILNSILS02725.1EA 0 EUR
SILNSILS02725.1 SILNSILS02725.5 SILNSILS02725.25
Bis(trimethylsilyl)methan
Bis(trimethylsilyl)methan
Formel: C₇H₂₀Si₂
MDL: MFCD00008275
CAS-Nummer: 2117-28-4
EINECS: 218-322-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR