(Brommethyl)trimethylsilan

Lieferant: CHEMPUR SILANE

SILS02925.5
SILNSILS02925.5EA 0 EUR
SILNSILS02925.5
(Brommethyl)trimethylsilan
(Bromomethyl)trimethylsilane
Formel: BrH₂CSi(CH₃)₃
Siedepunkt: 116 °C (1013 hPa)
Dichte: 1,17 g/cm³ (25 °C)
MDL: MFCD00000167
CAS-Nummer: 18243-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR