(4-Bromphenoxy)trimethylsilan

Lieferant: CHEMPUR SILANE

SILS02950.5 SILS02950.10 SILS02950.25
SILNSILS02950.5EA 0 EUR
SILNSILS02950.5 SILNSILS02950.10 SILNSILS02950.25
(4-Bromphenoxy)trimethylsilan
(4-Bromophenoxy)trimethylsilane
Formel: C₉H₁₃BrOSi
MDL: MFCD00040713
CAS-Nummer: 17878-44-3
EINECS: 241-831-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR