(4-Bromphenoxy)trimethylsilan
Lieferant: CHEMPUR SILANE
SILS02950.5
SILS02950.10
SILS02950.25
SILNSILS02950.5EA
0
EUR
SILNSILS02950.5
SILNSILS02950.10
SILNSILS02950.25
(4-Bromphenoxy)trimethylsilan
(4-Bromophenoxy)trimethylsilane
Formel:
C₉H₁₃BrOSi Molecular Weight: 245.20 g/mol |
MDL:
MFCD00040713 CAS-Nummer: 17878-44-3 EINECS: 241-831-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...