3-Brom-1-(trimethylsilyl)-1-propin
Lieferant: CHEMPUR SILANE
Synonyme:
3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
SILS03140.1
SILS03140.5
SILNSILS03140.1EA
0
EUR
SILNSILS03140.1
SILNSILS03140.5
3-Brom-1-(trimethylsilyl)-1-propin
3-Brom-1-(trimethylsilyl)-1-propin
Formel:
C₆H₁₁BrSi Molecular Weight: 191.14 g/mol |
MDL:
MFCD00134460 CAS-Nummer: 38002-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...