3-Brom-1-(trimethylsilyl)-1-propin

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane

SILS03140.1 SILS03140.5
SILNSILS03140.1EA 0 EUR
SILNSILS03140.1 SILNSILS03140.5
3-Brom-1-(trimethylsilyl)-1-propin
3-Brom-1-(trimethylsilyl)-1-propin
Formel: C₆H₁₁BrSi
Molecular Weight: 191.14 g/mol
MDL: MFCD00134460
CAS-Nummer: 38002-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR