Butyl(dimethyl)silan

Lieferant: CHEMPUR SILANE

SILS03315.10 SILS03315.25 SILS03315.100
SILNSILS03315.10EA 0 EUR
SILNSILS03315.10 SILNSILS03315.25 SILNSILS03315.100
Butyl(dimethyl)silan
Butyl(dimethyl)silan
Formel: C₆H₁₆Si
CAS-Nummer: 1001-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR