4-(tert-Butyldimethylsilyloxy)phenylboronsäure

Lieferant: CHEMPUR SILANE

Synonyme: 4-(tert-Butyl dimethylsiloxy)phenyl boronic acid, 4-(tert-Butyldimethylsiloxy)phenyl boronic acid

SILS03331.1
SILNSILS03331.1EA 0 EUR
SILNSILS03331.1
4-(tert-Butyldimethylsilyloxy)phenylboronsäure
4-(tert-Butyldimethylsilyloxy)phenylboronsäure
Formel: C₁₂H₂₁BO₃Si
Molecular Weight: 252,1900 g/mol
MDL: MFCD03093888
CAS-Nummer: 159191-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR