tert-Butylchlordiphenylsilan

Lieferant: CHEMPUR SILANE

Synonyme: tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane

SILS03387.10 S03387-25G SILS03387.100
SILNSILS03387.10EA 0 EUR
SILNSILS03387.10 FLUOS03387-25G SILNSILS03387.100
tert-Butylchlordiphenylsilan
tert-Butylchlordiphenylsilan
Formel: C₁₆H₁₉ClSi
MDL: MFCD00000497
CAS-Nummer: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR