tert-Butyltrichlorsilan

Lieferant: CHEMPUR SILANE

SILS03525.5 SILS03525.25
SILNSILS03525.5EA 0 EUR
SILNSILS03525.5 SILNSILS03525.25
tert-Butyltrichlorsilan
tert-Butyltrichlorsilan
Formel: C₄H₉Cl₃Si
MDL: MFCD00000480
CAS-Nummer: 18171-74-9
EINECS: 242-059-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR