Calciumdisilicid

Lieferant: CHEMPUR SILANE

Synonyme: Calcium silicide

SILS03575.50
SILNSILS03575.50EA 0 EUR
SILNSILS03575.50
Calciumdisilicid
Calcium disilicide
Formel: CaSi₂
Molecular Weight: 96.25 g/mol
MDL: MFCD00015977
CAS-Nummer: 12013-56-8
EINECS: 234-588-7
UN: 1405
ADR: 4.3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR