Dichlor(chlormethyl)methylsilan

Lieferant: CHEMPUR SILANE

SILS04175.25
SILNSILS04175.25EA 0 EUR
SILNSILS04175.25
Dichlor(chlormethyl)methylsilan
Dichlor(chlormethyl)methylsilan
Formel: C₂H₅Cl₃Si
Molecular Weight: 163,51 g/mol
MDL: MFCD00000874
CAS-Nummer: 1558-33-4
EINECS: 216-319-5
UN: 2988
ADR: 4.3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR