(Chlormethyl)trimethylsilan

Lieferant: CHEMPUR SILANE

SILS04425.10 SILS04425.25 SILS04425.100
SILNSILS04425.10EA 0 EUR
SILNSILS04425.10 SILNSILS04425.25 SILNSILS04425.100
(Chlormethyl)trimethylsilan
(Chloromethyl)trimethylsilane
Formel: ClH₂CSi(CH₃)₃
Molecular Weight: 122.67 g/mol
Siedepunkt: 98
Dichte: 0.8861 g/cm³ (20 °C)
MDL: MFCD00000878
CAS-Nummer: 2344-80-1
EINECS: 219-058-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR