(Chlormethyl)trimethylsilan
Lieferant: CHEMPUR SILANE
SILS04425.10
SILS04425.25
SILS04425.100
SILNSILS04425.10EA
0
EUR
SILNSILS04425.10
SILNSILS04425.25
SILNSILS04425.100
(Chlormethyl)trimethylsilan
(Chloromethyl)trimethylsilane
Formel:
ClH₂CSi(CH₃)₃ Molecular Weight: 122.67 g/mol Siedepunkt: 98 Dichte: 0.8861 g/cm³ (20 °C) |
MDL:
MFCD00000878 CAS-Nummer: 2344-80-1 EINECS: 219-058-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...