(3-Chlorpropyl)trichlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Trichloro(3-chloropropyl)silane

SILS04625.25 SILS04625.100
SILNSILS04625.25EA 0 EUR
SILNSILS04625.25 SILNSILS04625.100
(3-Chlorpropyl)trichlorsilan
(3-Chlorpropyl)trichlorsilan
Formel: ClH₂CCH₂CH₂SiCl₃
Molecular Weight: 211.98 g/mol
Siedepunkt: 176…177 °C (1013 hPa)
Dichte: 1.359 g/cm³ (20 °C)
MDL: MFCD00013694
CAS-Nummer: 2550-06-3
EINECS: 219-844-8
UN: 2987
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR