Cyclohexyl(dimethoxy)methylsilan

Lieferant: CHEMPUR SILANE

SILS05200.10 SILS05200.25
SILNSILS05200.10EA 0 EUR
SILNSILS05200.10 SILNSILS05200.25
Cyclohexyl(dimethoxy)methylsilan
Cyclohexyl(dimethoxy)methylsilane
Formel: C₉H₂₀O₂Si
MDL: MFCD00071752
CAS-Nummer: 17865-32-6
EINECS: 402-140-1
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR